Product Details
- SNP ID
-
rs11088420
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:38130692 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCAGAAAGGCTTTTGGCGACATC[A/C]CATTCACTTAGGAGCTGTGGTCCAT
- Phenotype
-
MIM: 604829
MIM: 613396
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DSCR4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116143076] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DSCR4
- Gene Name
- Down syndrome critical region 4
There are no transcripts associated with this gene.
- Gene
- DSCR8
- Gene Name
- Down syndrome critical region 8
There are no transcripts associated with this gene.
View Full Product Details