Product Details

SNP ID
rs11998184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:16994089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCAAATGTCAGTTCCCAATTT[C/T]ATTTTCAAATACCTAAAAAATATCC
Phenotype
MIM: 605558
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF20 PubMed Links

Gene Details

Gene
FGF20
Gene Name
fibroblast growth factor 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019851.2 Intron NP_062825.1

View Full Product Details