Product Details

SNP ID
rs7818801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:17865358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGTGCTCTACTACTGGCTTTCCT[C/G]GATAATTTGCACTCACACGTTCATT
Phenotype
MIM: 605776
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FGL1 PubMed Links

Gene Details

Gene
FGL1
Gene Name
fibrinogen like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004467.3 Intron NP_004458.3
NM_147203.2 Intron NP_671736.2
NM_201552.1 Intron NP_963846.1
NM_201553.1 Intron NP_963847.1
XM_017013232.1 Intron XP_016868721.1
XM_017013233.1 Intron XP_016868722.1

View Full Product Details