Product Details

SNP ID
rs2850020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33329895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATACTTCTTTTGAGGATTCTGG[A/G]AAGATGGCAGAGTAGAAAGCACCAG
Phenotype
MIM: 107450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNAR1 PubMed Links

Gene Details

Gene
IFNAR1
Gene Name
interferon alpha and beta receptor subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000629.2 Intron NP_000620.2
XM_005260964.2 Intron XP_005261021.1
XM_011529552.1 Intron XP_011527854.1

View Full Product Details