Product Details

SNP ID
rs2284553
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:33404389 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGGGCTCAGAACTGTCCGGGTCCC[A/G]TCAGTGTTGGGGCGGAAGAGGAAGA
Phenotype
MIM: 147569
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNGR2 PubMed Links

Gene Details

Gene
IFNGR2
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005534.3 Intron NP_005525.2
XM_005260969.2 Intron XP_005261026.1
XM_011529553.1 Intron XP_011527855.1
XM_011529554.2 Intron XP_011527856.1

View Full Product Details