Product Details

SNP ID
rs9974620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34725496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACTTTGGGGGCCTTGCCCCATC[C/T]AGAGGGGGCAGAACGTTTCAGTTGG
Phenotype
MIM: 615321
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLIC6 PubMed Links

Gene Details

Gene
CLIC6
Gene Name
chloride intracellular channel 6
There are no transcripts associated with this gene.

Gene
LINC00160
Gene Name
long intergenic non-protein coding RNA 160
There are no transcripts associated with this gene.

View Full Product Details