Product Details

SNP ID
rs35642400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34728447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAACCCTTCCAGGGGAAGTCGC[C/T]CTCATCAATACACAAGCCAGGGTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00160 PubMed Links

Gene Details

Gene
LINC00160
Gene Name
long intergenic non-protein coding RNA 160
There are no transcripts associated with this gene.

View Full Product Details