Product Details

SNP ID
rs261950
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:30301784 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCAGCTGAGAGTGAGTTCACACC[A/T]TGTACTCTAAAATGAGGAGGTGCTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HCG17 PubMed Links

Gene Details

Gene
HCG17
Gene Name
HLA complex group 17 (non-protein coding)
There are no transcripts associated with this gene.

Gene
HCG18
Gene Name
HLA complex group 18 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details