Product Details

SNP ID
rs13515
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:21761597 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATACAAATAGTGGAGGTGCTGTTC[C/T]GTGTCACTAATATGAATTAAGATGA
Phenotype
MIM: 176948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPK1 PubMed Links

Gene Details

Gene
MAPK1
Gene Name
mitogen-activated protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002745.4 3976 UTR 3 NP_002736.3
NM_138957.3 3976 Intron NP_620407.1

View Full Product Details