Product Details

SNP ID
rs3129881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32441707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCTCAGTCTTAATTTTTGGAGTA[C/T]TTCAGTCCCCAGGTATATTTTCCAT
Phenotype
MIM: 142860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HLA-DRA PubMed Links

Gene Details

Gene
HLA-DRA
Gene Name
major histocompatibility complex, class II, DR alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019111.4 Intron NP_061984.2

View Full Product Details