Product Details
- SNP ID
-
rs34841068
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45124096 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGGACAGCCCCAGCCCTGGGAGGCA[C/G]GTCTCCTTACCTTCTTCCAGTTCCT
- Phenotype
-
MIM: 609872
MIM: 605161
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
WFDC12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8124031] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- WFDC12
- Gene Name
- WAP four-disulfide core domain 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080869.1 |
|
Intron |
|
|
NP_543145.1 |
- Gene
- WFDC5
- Gene Name
- WAP four-disulfide core domain 5
There are no transcripts associated with this gene.
View Full Product Details