Product Details
- SNP ID
-
rs4499491
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45092778 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTCTCTCTGCTTGGAGTACTCCCC[A/C]CTGGAACCTCATTTGCTTAATTAGC
- Phenotype
-
MIM: 602905
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KCNS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8118000] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNS1
- Gene Name
- potassium voltage-gated channel modifier subfamily S member 1
View Full Product Details