Product Details

SNP ID
rs4828121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85019216 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGTGGGTGTTCCCTTCAATTTT[A/G]TATGTTGGAAACTAATACCGTATGT
Phenotype
MIM: 300955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOOL PubMed Links

Gene Details

Gene
APOOL
Gene Name
apolipoprotein O like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198450.5 Intron NP_940852.3
XM_011530859.2 Intron XP_011529161.1
XM_017029271.1 Intron XP_016884760.1
XM_017029272.1 Intron XP_016884761.1
XM_017029273.1 Intron XP_016884762.1

View Full Product Details