Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001162893.1 | 2431 | UTR 3 | NP_001156365.1 | ||
NM_001162894.1 | 2431 | UTR 3 | NP_001156366.1 | ||
NM_001162895.1 | 2431 | UTR 3 | NP_001156367.1 | ||
NM_001319230.1 | 2431 | UTR 3 | NP_001306159.1 | ||
NM_001319231.1 | 2431 | UTR 3 | NP_001306160.1 | ||
NM_014656.2 | 2431 | UTR 3 | NP_055471.2 | ||
XM_017002909.1 | 2431 | UTR 3 | XP_016858398.1 | ||
XM_017002910.1 | 2431 | UTR 3 | XP_016858399.1 | ||
XM_017002911.1 | 2431 | UTR 3 | XP_016858400.1 |