Product Details

SNP ID
rs11809587
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:175159308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGTCCTCACATACCTCAAGGCA[A/C]ATGTGTCACCTACATGTGCTCTCCA
Phenotype
MIM: 616696
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0040 PubMed Links

Gene Details

Gene
KIAA0040
Gene Name
KIAA0040
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162893.1 2530 UTR 3 NP_001156365.1
NM_001162894.1 2530 UTR 3 NP_001156366.1
NM_001162895.1 2530 UTR 3 NP_001156367.1
NM_001319230.1 2530 UTR 3 NP_001306159.1
NM_001319231.1 2530 UTR 3 NP_001306160.1
NM_014656.2 2530 UTR 3 NP_055471.2
XM_017002909.1 2530 UTR 3 XP_016858398.1
XM_017002910.1 2530 UTR 3 XP_016858399.1
XM_017002911.1 2530 UTR 3 XP_016858400.1

View Full Product Details