Product Details

SNP ID
rs2205845
Assay Type
Functionally Tested
NCBI dbSNP Submissions
58
Location
Chr.1:169796199 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCCCTTTTTTAGTTTTTGTGTAG[A/G]TACAGAACTGCCGTCTTCAAGGAGT
Phenotype
MIM: 615255
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C1orf112 PubMed Links

Gene Details

Gene
C1orf112
Gene Name
chromosome 1 open reading frame 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320047.1 Intron NP_001306976.1
NM_001320048.1 Intron NP_001306977.1
NM_001320050.1 Intron NP_001306979.1
NM_001320051.1 Intron NP_001306980.1
NM_018186.3 Intron NP_060656.2
XM_005245317.4 Intron XP_005245374.1
XM_011509735.2 Intron XP_011508037.1
XM_017001722.1 Intron XP_016857211.1
XM_017001723.1 Intron XP_016857212.1
XM_017001724.1 Intron XP_016857213.1
XM_017001725.1 Intron XP_016857214.1
Gene
METTL18
Gene Name
methyltransferase like 18
There are no transcripts associated with this gene.

View Full Product Details