Product Details
- SNP ID
-
rs10800478
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:169797157 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGACACATAGTAAGCATTCATTAAA[G/T]GGCTAAAAATATTTCATGTTTTAAA
- Phenotype
-
MIM: 615255
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf112
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79524498] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf112
- Gene Name
- chromosome 1 open reading frame 112
- Gene
- METTL18
- Gene Name
- methyltransferase like 18
There are no transcripts associated with this gene.
View Full Product Details