Product Details

SNP ID
rs17482426
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:167809159 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAATTTTATTTGATATTTGATGC[A/T]TGTGGTTAGAGGGGCTAAGACATCA
Phenotype
MIM: 605205
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADCY10 PubMed Links

Gene Details

Gene
ADCY10
Gene Name
adenylate cyclase 10, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167749.2 6829 Intron NP_001161221.1
NM_001297772.1 6829 Intron NP_001284701.1
NM_018417.5 6829 Intron NP_060887.2
XM_005245330.4 6829 Intron XP_005245387.1
XM_006711449.3 6829 UTR 3 XP_006711512.1
XM_011509760.2 6829 UTR 3 XP_011508062.1
XM_011509762.2 6829 UTR 3 XP_011508064.1
XM_011509763.2 6829 UTR 3 XP_011508065.1
XM_011509766.2 6829 Intron XP_011508068.1
XM_017001778.1 6829 Intron XP_016857267.1

View Full Product Details