Product Details

SNP ID
rs5960905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:57126526 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAGAGTGAACCTGAAAGGATAT[A/G]AGGATTAAGCTGCTGTATTCAGCTT
Phenotype
MIM: 300654 MIM: 300621 MIM: 300517
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAAH2 PubMed Links

Gene Details

Gene
FAAH2
Gene Name
fatty acid amide hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174912.3 Intron NP_777572.2
XM_005261982.1 Intron XP_005262039.1
XM_005261985.2 Intron XP_005262042.1
XM_005261986.3 Intron XP_005262043.1
XM_005261987.3 Intron XP_005262044.1
XM_011530767.2 Intron XP_011529069.1
XM_017029290.1 Intron XP_016884779.1
XM_017029291.1 Intron XP_016884780.1
XM_017029292.1 Intron XP_016884781.1
XM_017029293.1 Intron XP_016884782.1
XM_017029294.1 Intron XP_016884783.1
Gene
SPIN2A
Gene Name
spindlin family member 2A
There are no transcripts associated with this gene.

Gene
SPIN2B
Gene Name
spindlin family member 2B
There are no transcripts associated with this gene.

View Full Product Details