Product Details
- SNP ID
-
rs2235140
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38865130 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTTCAGCATCCTCCCCAAACACC[C/G]GGAGGAAGAGGGAGCTTGGAATGGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CBX6
PubMed Links
Gene Details
- Gene
- CBX6
- Gene Name
- chromobox 6
View Full Product Details