Product Details

SNP ID
rs8116218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45867691 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACATTGCTATCAACTGGGAAGAGT[A/C]TGTGACTTTATGCCCAGTTTCCCCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZSWIM3 PubMed Links

Gene Details

Gene
ZSWIM3
Gene Name
zinc finger SWIM-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080752.3 Intron NP_542790.2

View Full Product Details