Product Details

SNP ID
rs6094216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45837600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTGGCATTATAGGTTAATCCA[C/T]ATTCAGGTTCTCAGACCTGGGAGGC
Phenotype
MIM: 608123 MIM: 191039
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOT8 PubMed Links

Gene Details

Gene
ACOT8
Gene Name
acyl-CoA thioesterase 8
There are no transcripts associated with this gene.

Gene
SNX21
Gene Name
sorting nexin family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042632.2 Intron NP_001036097.1
NM_001042633.2 Intron NP_001036098.1
NM_033421.3 Intron NP_219489.1
NM_152897.2 Intron NP_690857.1
XM_005260608.4 Intron XP_005260665.1
XM_011529095.2 Intron XP_011527397.1
XM_011529096.2 Intron XP_011527398.1
XM_011529098.2 Intron XP_011527400.1
XM_011529099.2 Intron XP_011527401.1
XM_017028120.1 Intron XP_016883609.1
Gene
TNNC2
Gene Name
troponin C2, fast skeletal type
There are no transcripts associated with this gene.

View Full Product Details