Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001198993.1 | Intron | NP_001185922.1 | |||
NM_001198994.1 | Intron | NP_001185923.1 | |||
NM_001198995.1 | Intron | NP_001185924.1 | |||
NM_023018.4 | Intron | NP_075394.3 | |||
XM_005244778.2 | Intron | XP_005244835.1 | |||
XM_006710837.2 | Intron | XP_006710900.1 | |||
XM_006710838.2 | Intron | XP_006710901.1 | |||
XM_006710839.1 | Intron | XP_006710902.1 | |||
XM_011542006.1 | Intron | XP_011540308.1 | |||
XM_011542007.1 | Intron | XP_011540309.1 | |||
XM_011542008.1 | Intron | XP_011540310.1 | |||
XM_011542009.2 | Intron | XP_011540311.1 | |||
XM_017002143.1 | Intron | XP_016857632.1 | |||
XM_017002144.1 | Intron | XP_016857633.1 | |||
XM_017002145.1 | Intron | XP_016857634.1 | |||
XM_017002146.1 | Intron | XP_016857635.1 |