Product Details

SNP ID
rs6002171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40827855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAGACTAATCTCGCCTTAAAA[A/G]CAAAGAGTCATCTAACAGTTCCTTA
Phenotype
MIM: 606795 MIM: 606796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A17 PubMed Links

Gene Details

Gene
SLC25A17
Gene Name
solute carrier family 25 member 17
There are no transcripts associated with this gene.

Gene
ST13
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278589.1 Intron NP_001265518.1
NM_003932.4 Intron NP_003923.2

View Full Product Details