Product Details
- SNP ID
-
rs5755921
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:35708683 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAGATAAATATACGCTTTCCTCTG[C/T]GCTGATTTTTTTCTGATGAGAAGAC
- Phenotype
-
MIM: 607255
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APOL5
PubMed Links
Gene Details
- Gene
- APOL5
- Gene Name
- apolipoprotein L5
View Full Product Details