Product Details
- SNP ID
-
rs11247685
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
35
- Location
-
Chr.1:27783761 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATAGGAAATACCCAATTATTTTCCT[C/T]CAAAAAGTTGTAGCACATCAGTCTA
- Phenotype
-
MIM: 606892
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
STX12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112485554] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- STX12
- Gene Name
- syntaxin 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177424.2 |
|
Intron |
|
|
NP_803173.1 |
View Full Product Details