Product Details

SNP ID
rs2267103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26610225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAAGATATGGAAACTGAGGCTAA[A/G]AGGGGTTAAGTCCCCAGCCTAAGGT
Phenotype
MIM: 123631 MIM: 600929
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYBA4 PubMed Links

Gene Details

Gene
CRYBA4
Gene Name
crystallin beta A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001886.2 Intron NP_001877.1
XM_006724140.3 Intron XP_006724203.1
XM_017028598.1 Intron XP_016884087.1
Gene
CRYBB1
Gene Name
crystallin beta B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001887.3 Intron NP_001878.1
XM_011529899.2 Intron XP_011528201.1

View Full Product Details