Product Details

SNP ID
rs34347370
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:175320867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGAATTTTCATTACCTAATTGT[A/G]CTGTCCCACCATGCCAATTTCTACT
Phenotype
MIM: 601995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNR PubMed Links

Gene Details

Gene
TNR
Gene Name
tenascin R
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003285.2 6731 Intron NP_003276.3
XM_011509949.2 6731 UTR 3 XP_011508251.1
XM_017002218.1 6731 UTR 3 XP_016857707.1
XM_017002219.1 6731 UTR 3 XP_016857708.1

View Full Product Details