Product Details
- SNP ID
-
rs11208725
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:40702209 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATTTGTCATCTCCTTAAGGTCC[A/G]TTTATTAGTTTATTTCATTCCTTCA
- Phenotype
-
MIM: 605344
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NFYC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74888385] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NFYC
- Gene Name
- nuclear transcription factor Y subunit gamma
- Gene
- NFYC-AS1
- Gene Name
- NFYC antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details