Product Details

SNP ID
rs503706
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:170665943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGAATGAAGAACCAGGTGTCC[C/T]CAAGTTCCGCATAAAGGGCTAGGCG
Phenotype
MIM: 167420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRRX1 PubMed Links

Gene Details

Gene
PRRX1
Gene Name
paired related homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006902.4 Intron NP_008833.1
NM_022716.3 Intron NP_073207.1
XM_006711388.3 Intron XP_006711451.1

View Full Product Details