Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001242894.1 | 779 | Intron | NP_001229823.1 | ||
NM_001242895.1 | 779 | Intron | NP_001229824.1 | ||
NM_024493.3 | 779 | Intron | NP_077819.2 | ||
XM_005249423.3 | 779 | Intron | XP_005249480.1 | ||
XM_006715215.2 | 779 | Intron | XP_006715278.1 | ||
XM_006715216.2 | 779 | Intron | XP_006715279.1 | ||
XM_006715218.3 | 779 | Intron | XP_006715281.1 | ||
XM_017011320.1 | 779 | Intron | XP_016866809.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135215.1 | 779 | Intron | NP_001128687.1 | ||
NM_001135216.1 | 779 | Intron | NP_001128688.1 | ||
NM_001243241.1 | 779 | Intron | NP_001230170.1 | ||
NM_001243242.1 | 779 | Intron | NP_001230171.1 | ||
NM_001243243.1 | 779 | Intron | NP_001230172.1 | ||
NM_001243244.1 | 779 | Intron | NP_001230173.1 | ||
NM_030899.4 | 779 | Intron | NP_112161.3 | ||
NM_145909.2 | 779 | Intron | NP_665916.1 | ||
XM_005249295.1 | 779 | Intron | XP_005249352.1 | ||
XM_005249296.4 | 779 | UTR 5 | XP_005249353.1 | ||
XM_011514807.2 | 779 | UTR 5 | XP_011513109.1 | ||
XM_011514808.2 | 779 | UTR 5 | XP_011513110.1 | ||
XM_011514809.1 | 779 | Intron | XP_011513111.1 | ||
XM_011514811.2 | 779 | Intron | XP_011513113.1 | ||
XM_011514812.2 | 779 | Intron | XP_011513114.1 | ||
XM_011514813.2 | 779 | Intron | XP_011513115.1 | ||
XM_017011196.1 | 779 | UTR 5 | XP_016866685.1 |