Product Details

SNP ID
rs2038256
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:28780143 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACTGGAGTTGAGGAATTTCAGGAG[G/T]CACTCAAAGAACTGCATCCATTTCT
Phenotype
MIM: 164874
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FOXG1 PubMed Links

Gene Details

Gene
FOXG1
Gene Name
forkhead box G1
There are no transcripts associated with this gene.

Gene
LINC01551
Gene Name
long intergenic non-protein coding RNA 1551
There are no transcripts associated with this gene.

Gene
LOC105370424
Gene Name
uncharacterized LOC105370424
There are no transcripts associated with this gene.

View Full Product Details