Product Details

SNP ID
rs2269505
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:50560517 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGGACCACGGACCCTCAGGAAGCA[A/G]AGGAACCTGGGGTCCTAGCCGCCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf18 PubMed Links

Gene Details

Gene
C3orf18
Gene Name
chromosome 3 open reading frame 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171740.2 Intron NP_001165211.1
NM_001171741.2 Intron NP_001165212.1
NM_001171743.2 Intron NP_001165214.2
NM_016210.4 Intron NP_057294.2
XM_011533781.2 Intron XP_011532083.1
XM_011533782.2 Intron XP_011532084.1
XM_011533783.2 Intron XP_011532085.1
XM_011533784.2 Intron XP_011532086.1
XM_011533785.2 Intron XP_011532087.1
XM_011533788.1 Intron XP_011532090.1
XM_011533789.1 Intron XP_011532091.1
XM_011533790.1 Intron XP_011532092.1
XM_017006545.1 Intron XP_016862034.1
XM_017006546.1 Intron XP_016862035.1
XM_017006547.1 Intron XP_016862036.1
Gene
HEMK1
Gene Name
HemK methyltransferase family member 1
There are no transcripts associated with this gene.

View Full Product Details