Product Details

SNP ID
rs3731586
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:31523958 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCTACCCTATATTCAGATTAAGAAG[C/T]TCCACGATCATGCTAACTTGAAAGG
Phenotype
MIM: 607306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRD5A2 PubMed Links

Gene Details

Gene
SRD5A2
Gene Name
steroid 5 alpha-reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000348.3 3789 Intron NP_000339.2
XM_011533069.2 3789 UTR 3 XP_011531371.1
XM_011533072.2 3789 UTR 3 XP_011531374.1

View Full Product Details