Product Details

SNP ID
rs4952073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31235115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCAAGTGTAAAGCCATTCCCAG[C/T]CTCCCCATCTTATGGATGATGAAAC
Phenotype
MIM: 610229 MIM: 605891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPN14 PubMed Links

Gene Details

Gene
CAPN14
Gene Name
calpain 14
There are no transcripts associated with this gene.

Gene
EHD3
Gene Name
EH domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014600.2 Intron NP_055415.1
XM_011532806.2 Intron XP_011531108.1

View Full Product Details