Product Details
- SNP ID
-
rs6007806
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:46296342 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGCTCCCCAGCCGGCCTGTAAAACC[C/G]AGGACAGGCTCCCGCCACGCCCTCG
- Phenotype
-
MIM: 607477
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GTSE1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7285629] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTSE1
- Gene Name
- G2 and S-phase expressed 1
- Gene
- GTSE1-AS1
- Gene Name
- GTSE1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
- Gene
- TTC38
- Gene Name
- tetratricopeptide repeat domain 38
There are no transcripts associated with this gene.
View Full Product Details