Product Details

SNP ID
rs2397084
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52237046 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCTTCCTCCGGACGACCAGGGTC[C/T]CTTGCTGGATGGGAACGGAATTCAT
Phenotype
MIM: 606496
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
IL17F PubMed Links

Gene Details

Gene
IL17F
Gene Name
interleukin 17F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052872.3 514 Missense Mutation GAG,GGG E126G NP_443104.1
XM_011514276.1 514 Missense Mutation GAG,GGG E126G XP_011512578.1

View Full Product Details