Product Details

SNP ID
rs4820379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39508608 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTCTATCTCAGTGGCCACTGGAG[A/G]GAGGGTGTCTGCTGTTCTTCCCTGA
Phenotype
MIM: 615497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIEF1 PubMed Links

Gene Details

Gene
MIEF1
Gene Name
mitochondrial elongation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304564.1 Intron NP_001291493.1
NM_019008.5 Intron NP_061881.2
XM_011530236.1 Intron XP_011528538.1
XM_011530237.1 Intron XP_011528539.1
XM_011530238.1 Intron XP_011528540.1
XM_017028838.1 Intron XP_016884327.1

View Full Product Details