Product Details

SNP ID
rs9496878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:144144082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCTTCAACTCAGTGCTTACTAC[A/G]GTCATTCATTAACTCATCTTTCAAC
Phenotype
MIM: 605014
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STX11 PubMed Links

Gene Details

Gene
STX11
Gene Name
syntaxin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003764.3 Intron NP_003755.2
XM_011536213.2 Intron XP_011534515.1
XM_011536214.2 Intron XP_011534516.1
XM_011536217.2 Intron XP_011534519.1
XM_011536218.2 Intron XP_011534520.1
XM_017011400.1 Intron XP_016866889.1

View Full Product Details