Product Details
- SNP ID
-
rs3484
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:36800827 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CTGAACAGAAATGCAGGAGGGTGGC[A/G]AGAGGGGCCGAGATTGGGTGTTCAG
- Phenotype
-
MIM: 168890
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PVALB
PubMed Links
Gene Details
- Gene
- PVALB
- Gene Name
- parvalbumin
View Full Product Details