Product Details

SNP ID
rs2001063
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:36800927 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACCAGAGTGGAGAATTCTGCAGAA[A/C]AAAATGACAAGGAAAGTGAGTCAGA
Phenotype
MIM: 168890
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
PVALB PubMed Links
Additional Information
For this assay, SNP(s) [rs78079590] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PVALB
Gene Name
parvalbumin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001315532.1 Intron NP_001302461.1
NM_002854.2 Intron NP_002845.1

View Full Product Details