Product Details

SNP ID
rs133072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40679539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTACGGAGGAAGACCCCCTTCCC[A/G]ACTGCGGGGCTTGCGCTCCGGGACA
Phenotype
MIM: 601751
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCHR1 PubMed Links

Gene Details

Gene
MCHR1
Gene Name
melanin concentrating hormone receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005297.3 362 Missense Mutation AAC,GAC N32D NP_005288.3

View Full Product Details