Product Details

SNP ID
rs11970734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10756075 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACTCTGATCATAGTCGTGATGCT[C/G]CCAGTATAATATTTTATACCTGAAG
Phenotype
MIM: 154235
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAK PubMed Links

Gene Details

Gene
MAK
Gene Name
male germ cell associated kinase
There are no transcripts associated with this gene.

Gene
TMEM14B
Gene Name
transmembrane protein 14B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127711.2 Intron NP_001121183.1
NM_001286484.1 Intron NP_001273413.1
NM_001286488.1 Intron NP_001273417.1
NM_001286489.1 Intron NP_001273418.1
NM_030969.4 Intron NP_112231.3

View Full Product Details