Product Details
- SNP ID
-
rs126405
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:10764494 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTGGCCACCCAGTCTGTCCTCCC[A/G]TGCACTGAGGGAATGGGCTGTGCAC
- Phenotype
-
MIM: 154235
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAK
PubMed Links
Gene Details
- Gene
- MAK
- Gene Name
- male germ cell associated kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242385.1 |
1229 |
Silent Mutation |
CAC,CAT |
H570H |
NP_001229314.1 |
NM_001242957.2 |
1229 |
Silent Mutation |
CAC,CAT |
H635H |
NP_001229886.1 |
NM_005906.5 |
1229 |
Silent Mutation |
CAC,CAT |
H610H |
NP_005897.1 |
XM_011514619.2 |
1229 |
Silent Mutation |
CAC,CAT |
H635H |
XP_011512921.1 |
XM_011514620.2 |
1229 |
Silent Mutation |
CAC,CAT |
H635H |
XP_011512922.1 |
XM_011514621.2 |
1229 |
Silent Mutation |
CAC,CAT |
H616H |
XP_011512923.1 |
XM_011514622.2 |
1229 |
Silent Mutation |
CAC,CAT |
H570H |
XP_011512924.1 |
XM_011514624.2 |
1229 |
Silent Mutation |
CAC,CAT |
H375H |
XP_011512926.1 |
XM_017010863.1 |
1229 |
Silent Mutation |
CAC,CAT |
H635H |
XP_016866352.1 |
XM_017010864.1 |
1229 |
Silent Mutation |
CAC,CAT |
H610H |
XP_016866353.1 |
XM_017010865.1 |
1229 |
Silent Mutation |
CAC,CAT |
H551H |
XP_016866354.1 |
XM_017010866.1 |
1229 |
Intron |
|
|
XP_016866355.1 |
- Gene
- TMEM14B
- Gene Name
- transmembrane protein 14B
There are no transcripts associated with this gene.
View Full Product Details