Product Details
- SNP ID
-
rs12133933
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
38
- Location
-
Chr.1:37492788 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTTAGACCCCAAATCTTATCCCTT[A/G]GACCAATCATAACATAGGATGTAAA
- Phenotype
-
MIM: 611001
MIM: 610562
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MEAF6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114745536] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MEAF6
- Gene Name
- MYST/Esa1 associated factor 6
- Gene
- MIR5581
- Gene Name
- microRNA 5581
There are no transcripts associated with this gene.
- Gene
- ZC3H12A
- Gene Name
- zinc finger CCCH-type containing 12A
There are no transcripts associated with this gene.
View Full Product Details