Product Details

SNP ID
rs7515306
Assay Type
Validated
NCBI dbSNP Submissions
66
Location
Chr.1:37534584 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTGCTGGCACGTTGTTTGTGTCAC[A/G]TTTAGATTCCGATTAAATGACATCA
Phenotype
MIM: 608241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNIP1 PubMed Links

Gene Details

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024700.3 4470 UTR 3 NP_078976.2

View Full Product Details