Product Details

SNP ID
rs847080
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:46541222 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGATCGCCAACCAGCCTGGAGGAAC[A/C]ATGAGCTCTCTCTGCACTCCTTGTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OCSTAMP PubMed Links

Gene Details

Gene
OCSTAMP
Gene Name
osteoclast stimulatory transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080721.2 1489 UTR 3 NP_542452.1
XM_011528532.2 1489 UTR 3 XP_011526834.1

View Full Product Details