Product Details

SNP ID
rs12125682
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:175016421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGTTTACAAATGAGGAAACTG[C/G]GGCAAAGAGAAATGAGAGTAGGAAA
Phenotype
MIM: 606186 MIM: 611978
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CACYBP PubMed Links

Gene Details

Gene
CACYBP
Gene Name
calcyclin binding protein
There are no transcripts associated with this gene.

Gene
MRPS14
Gene Name
mitochondrial ribosomal protein S14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022100.2 Intron NP_071383.1

View Full Product Details