Product Details

SNP ID
rs6527864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18686253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTAAGATTTGATCCAGAGACT[A/T]AGGATGGAGGGCTCCGGCTCCATTT
Phenotype
MIM: 300109
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PPEF1 PubMed Links

Gene Details

Gene
PPEF1
Gene Name
protein phosphatase with EF-hand domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006240.2 Intron NP_006231.2
NM_152224.1 Intron NP_689410.1
NM_152226.1 Intron NP_689412.1
XM_005274553.2 Intron XP_005274610.1
XM_017029612.1 Intron XP_016885101.1
Gene
PPEF1-AS1
Gene Name
PPEF1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details