Product Details

SNP ID
rs4820324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38203850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGGGGGAAGCTCTGGAAGTCT[C/G]TGCAGACAGCGTATTTTCCAGGTGA
Phenotype
MIM: 604877
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAFF PubMed Links

Gene Details

Gene
MAFF
Gene Name
MAF bZIP transcription factor F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161572.1 Intron NP_001155044.1
NM_001161573.1 Intron NP_001155045.1
NM_001161574.1 Intron NP_001155046.1
NM_012323.3 Intron NP_036455.1

View Full Product Details